Novus Biologicals
Manufacturer Code:NBP131215
Catalog # NBP131215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PIASy. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 SUMO-protein ligase PIAS4; E3 SUMO-protein ligase PIAS4 FLJ12419 MGC35296 Piasg PIAS-gamma PIASy Protein inhibitor of activated STAT protein 4 Protein inhibitor of activated STAT protein gamma protein inhibitor of activated STAT protein PIASy protein inhibitor of activated STAT 4 zinc finger MIZ-type containing 6 ZMIZ6; PIAS-gamma; PIASy; Protein inhibitor of activated STAT protein 4; Protein inhibitor of activated STAT protein gamma; protein inhibitor of activated STAT protein PIASy; protein inhibitor of activated STAT, 4; RING-type E3 ubiquitin transferase PIAS4; zinc finger, MIZ-type containing 6
Gene Aliases: PIAS-gamma; PIAS4; PIASG; PIASY; ZMIZ6
UniProt ID: (Human) Q8N2W9
Entrez Gene ID: (Human) 51588
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.