R&D Systems
Manufacturer Code:AF5120SP
Catalog # AF5120SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PIAS3 Met10-Asp628 Accession # Q9Y6X2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 SUMO-protein ligase PIAS3; E3 SUMO-protein transferase PIAS3; MIZ-type containing 5 Protein inhibitor of activated STAT protein 3 protein inhibitor of activated STAT 3 ZMIZ5; Protein inhibitor of activated STAT protein 3; protein inhibitor of activated STAT, 3; zinc finger, MIZ-type containing 5
Gene Aliases: PIAS3; ZMIZ5
UniProt ID: (Human) Q9Y6X2
Entrez Gene ID: (Human) 10401
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.