Abnova
Manufacturer Code:H00010401A01
Catalog # 89-106-303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 453 a.a. &bsim 550 a.a) partial recombinant protein with GST tag.,PIAS3 (NP_006090 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PIAS3.
For Research Use Only
Protein Aliases: E3 SUMO-protein ligase PIAS3; E3 SUMO-protein transferase PIAS3; FLJ14651; Protein inhibitor of activated STAT protein 3; protein inhibitor of activated STAT, 3; zinc finger, MIZ-type containing 5; ZMIZ5
Gene Aliases: PIAS3; ZMIZ5
UniProt ID: (Human) Q9Y6X2
Entrez Gene ID: (Human) 10401
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.