Proteintech
Manufacturer Code:134861AP150UL
Catalog # 50-172-8778
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PIAS3 Fusion Protein Ag4304 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 SUMO protein ligase PIAS3 PIAS3 ZMIZ5; E3 SUMO-protein ligase PIAS3; E3 SUMO-protein transferase PIAS3; KChAP; potassium channel regulatory protein KChAP; Potassium channel-associated protein; Protein inhibitor of activated STAT protein 3; protein inhibitor of activated STAT, 3; zinc finger, MIZ-type containing 5
Gene Aliases: PIAS3; Pias3l; ZMIZ5
UniProt ID: (Human) Q9Y6X2, (Mouse) O54714, (Rat) O70260
Entrez Gene ID: (Human) 10401, (Mouse) 229615, (Rat) 83614
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.