Abnova
Manufacturer Code:H00010401K
Catalog # 89-201-426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PIAS3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PIAS3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: E3 SUMO-protein ligase PIAS3; E3 SUMO-protein transferase PIAS3; FLJ14651 ZMIZ5; Protein inhibitor of activated STAT protein 3; protein inhibitor of activated STAT, 3; zinc finger, MIZ-type containing 5
Gene Aliases: PIAS3; ZMIZ5
UniProt ID: (Human) Q9Y6X2
Entrez Gene ID: (Human) 10401
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.