Invitrogen
Manufacturer Code:PA520952
Catalog # PIPA520952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the amino terminus of human PIAS2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Androgen receptor-interacting protein 3; ARIP3; ARIP3 DIP MIZ MIZ1 PIASX PIASX-ALPHA PIASX-BETA SIZ2 ZMIZ4 DAB2-interacting protein E3 SUMO-protein ligase PIAS2 androgen receptor-interacting protein 3 msx-interacting zinc finger protein protein inhibitor of activated STAT X zinc finger MIZ-type containing 4; DAB2-interacting protein; DIP; E3 SUMO-protein ligase PIAS2; E3 SUMO-protein transferase PIAS2; Miz1; Msx-interacting zinc finger protein; Msx-interacting-zinc finger; Msx-interacting-zinc finger protein; PIAS-NY protein; PIASxalpha6; Protein inhibitor of activated STAT x; Protein inhibitor of activated STAT2; RING-type E3 ubiquitin transferase PIAS2; zinc finger, MIZ-type containing 4
Gene Aliases: 6330408K17Rik; AI462206; ARIP3; AU018068; Dib; DIP; MIZ; MIZ1; PIAS2; PIASX; PIASX-ALPHA; PIASX-BETA; PIASxalpha; PIASxb; PIASxbeta; SIZ2; ZMIZ4
UniProt ID: (Human) O75928, (Rat) Q6AZ28, (Mouse) Q8C5D8
Entrez Gene ID: (Human) 9063, (Rat) 83422, (Mouse) 17344
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.