Novus Biologicals
Manufacturer Code:NBP219819
Catalog # NBP219819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human PIAS2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Androgen receptor-interacting protein 3; Androgen receptor-interacting protein 3 ARIP3 DAB2-interacting protein DIP E3 SUMO-protein ligase PIAS2 MGC102682 miz Miz1 Msx-interacting zinc finger protein Msx-interacting-zinc finger PIAS-NY protein PIASX PIASX-ALPHA PIASX-BETA Protein inhibitor of activated STAT x protein inhibitor of activated STAT 2 Protein inhibitor of activated STAT2 SIZ2 zinc finger MIZ-type containing 4 ZMIZ4; ARIP3; DAB2-interacting protein; DIP; E3 SUMO-protein ligase PIAS2; E3 SUMO-protein transferase PIAS2; Miz1; Msx-interacting zinc finger protein; PIAS-NY protein; Protein inhibitor of activated STAT x; Protein inhibitor of activated STAT2; zinc finger, MIZ-type containing 4
Gene Aliases: ARIP3; DIP; MIZ; MIZ1; PIAS2; PIASX; PIASX-ALPHA; PIASX-BETA; SIZ2; ZMIZ4
UniProt ID: (Human) O75928
Entrez Gene ID: (Human) 9063
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.