Invitrogen
Manufacturer Code:MA535571
Catalog # PIMA535571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1116 |
Immunogen | A synthesized peptide derived from human PIAS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2900068C24Rik DDXBP1 DEAD/H box-binding protein 1 E3 SUMO-protein ligase PIAS1 E3 SUMO-protein transferase PIAS1 GBP Gu binding protein GU/RH-II Gu-binding protein Pias1 Protein inhibitor of activated STAT protein 1 RING-type E3 ubiquitin transferase PIAS1 RNA helicase II-binding protein ZMIZ3; AR interacting protein; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein transferase PIAS1; GBP; Gu-binding protein; Protein inhibitor of activated STAT protein 1; protein inhibitor of activated STAT-1; RING-type E3 ubiquitin transferase PIAS1; RNA helicase II-binding protein; zinc finger, MIZ-type containing 3
Gene Aliases: 2900068C24Rik; DDXBP1; GBP; GU/RH-II; PIAS1; ZMIZ3
UniProt ID: (Human) Q99751, (Mouse) O88907
Entrez Gene ID: (Human) 8554, (Mouse) 56469
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.