Novus Biologicals
Manufacturer Code:NBP29455820UL
Catalog # NB023641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 422-651 of human PIAS1 (NP_057250.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AR interacting protein; AR interacting protein DDXBP1MGC141878 DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1 DEAD/H box-binding protein 1 E3 SUMO-protein ligase PIAS1 GBPRNA helicase II-binding protein GU/RH-II Gu-binding protein MGC141879 Protein inhibitor of activated STAT protein 1 protein inhibitor of activated STAT 1 protein inhibitor of activated STAT-1 zinc finger MIZ-type containing 3 ZMIZ3; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein transferase PIAS1; GBP; Gu-binding protein; Protein inhibitor of activated STAT protein 1; protein inhibitor of activated STAT-1; RNA helicase II-binding protein; zinc finger, MIZ-type containing 3
Gene Aliases: DDXBP1; GBP; GU/RH-II; PIAS1; ZMIZ3
UniProt ID: (Human) Q99751
Entrez Gene ID: (Human) 8554
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.