Abnova
Manufacturer Code:PAB19242
Catalog # 89-120-773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acids near C-terminus of human PIAS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of PIAS1.
For Research Use Only
Protein Aliases: AR interacting protein; DDXBP1; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein transferase PIAS1; GBP; Gu-binding protein; GU/RH-II; MGC141878; MGC141879; Protein inhibitor of activated STAT protein 1; protein inhibitor of activated STAT-1; RNA helicase II-binding protein; zinc finger, MIZ-type containing 3; ZMIZ3
Gene Aliases: DDXBP1; GBP; GU/RH-II; PIAS1; ZMIZ3
UniProt ID: (Human) Q99751
Entrez Gene ID: (Human) 8554
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.