Proteintech
Manufacturer Code:124111AP150UL
Catalog # 50-172-7964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PI4KA Fusion Protein Ag3062 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: leuserpin 2; phosphatidylinositol 4-kinase 230; phosphatidylinositol 4-kinase a; Phosphatidylinositol 4-kinase alpha; Phosphatidylinositol 4-Kinase III alpha; phosphatidylinositol 4-kinase, catalytic, alpha; phosphatidylinositol 4-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4-kinase, type III, alpha; PI4 kinase alpha PI4K ALPHA pi4K230 PI4KA PIK4 PIK4CA PtdIns 4 kinase alpha; PI4-kinase alpha; PI4K-alpha; ptdIns-4-kinase alpha; testicular secretory protein Li 35
Gene Aliases: PI4K-ALPHA; pi4K230; PI4KA; PIK4; PIK4CA; PMGYCHA
UniProt ID: (Human) P42356, (Rat) O08662, (Mouse) E9Q3L2
Entrez Gene ID: (Human) 5297, (Rat) 64161, (Mouse) 224020
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.