Novus Biologicals
Manufacturer Code:NBP267504
Catalog # NBP267504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ST04-77 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: P85B phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta phosphatidylinositol 3-kinase regulatory subunit beta phosphatidylinositol 3-kinase regulatory subunit polypeptide 2 (p85 beta) phosphoinositide-3-kinase regulatory subunit 2 (beta) phosphoinositide-3-kinase regulatory subunit 2 (p85 beta) phosphoinositide-3-kinase regulatory subunit polypeptide 2 (p85 beta) PI3K regulatory subunit beta PI3-kinase regulatory subunit beta PI3-kinase subunit p85-beta ptdIns-3-kinase regulatory subunit beta ptdIns-3-kinase regulatory subunit p85-beta; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; Phosphatidylinositol 3-kinase regulatory subunit beta; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 2 (beta); PI3-kinase regulatory subunit beta; PI3-kinase subunit p85-beta; PI3K regulatory subunit beta; ptdIns-3-kinase regulatory subunit p85-beta
Gene Aliases: MPPH; MPPH1; p85; p85-BETA; P85B; PIK3R2
UniProt ID: (Human) O00459
Entrez Gene ID: (Human) 5296
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.