Novus Biologicals
Manufacturer Code:NBP219815
Catalog # NBP219815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PI3 kinase p85 alpha. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GRB1PI3-kinase subunit p85-alpha p85 p85-ALPHA Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha phosphatidylinositol 3-kinase regulatory subunit alpha phosphatidylinositol 3-kinase regulatory subunit polypeptide 1 (p85 alpha) phosphatidylinositol 3-kinase regulatory 1 phosphatidylinositol 3-kinase-associated p-85 alpha phosphoinositide-3-kinase regulatory subunit 1 (alpha) PI3K regulatory subunit alpha PI3-kinase regulatory subunit alpha PtdIns-3-kinase regulatory subunit alpha PtdIns-3-kinase regulatory subunit p85-alpha; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; Phosphatidylinositol 3-kinase regulatory subunit alpha; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphatidylinositol 3-kinase-associated p-85 alpha; phosphoinositide-3-kinase regulatory subunit; phosphoinositide-3-kinase regulatory subunit alpha; phosphoinositide-3-kinase, regulatory subunit 1 (alpha); PI3-kinase regulatory subunit alpha; PI3-kinase subunit p85-alpha; PI3K regulatory subunit alpha; ptdIns-3-kinase regulatory subunit alpha
Gene Aliases: AGM7; GRB1; IMD36; p85; p85-ALPHA; PIK3R1
UniProt ID: (Human) P27986
Entrez Gene ID: (Human) 5295
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.