Novus Biologicals
Manufacturer Code:NBP273397AF594
Catalog # NP273397A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B1 |
Immunogen | Full length recombinant protein of human PI 3-Kinase p110 gamma (NP_002640) produced in HEK293T cells. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 1-phosphatidylinositol 3-kinase; 1-phosphatidylinositol 3-kinase EC 2.7.1 EC 2.7.1.153 p110-gamma p120-PI3K phosphatidylinositol 3 kinase gamma p110 gamma phosphatidylinositol 3-kinase catalytic 110-kD gamma phosphatidylinositol 3-kinase catalytic gamma polypeptide Phosphatidylinositol-45-bisphosphate 3-kinase 110 kDa catalytic subunit gamma phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit gamma isoform phosphoinositide-3-kinase gamma catalytic subunit phosphoinositide-3-kinase catalytic gamma polypeptide PI3CG PI3K PI3Kgamma PI3K-gamma PI3-kinase subunit gamma PIK3 PtdIns-3-kinase subunit gamma PtdIns-3-kinase subunit p110-gamma; p120-PI3K; phosphatidylinositol 3 kinase gamma, p110 gamma; phosphatidylinositol 3-kinase catalytic 110-kD gamma; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit gamma isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit gamma; Phosphoinositide-3-kinase catalytic gamma polypeptide; phosphoinositide-3-kinase gamma catalytic subunit; PI3-kinase subunit gamma; ptdIns-3-kinase subunit gamma; PtdIns-3-kinase subunit p110-gamma; Serine/threonine protein kinase PIK3CG
Gene Aliases: p110gamma; p120-PI3K; PI3CG; PI3K; PI3Kgamma; PIK3; PIK3CG
UniProt ID: (Human) P48736
Entrez Gene ID: (Human) 5294
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.