Novus Biologicals
Manufacturer Code:NBP273396
Catalog # NBP273396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D11 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 286-610 of human PIK3CD (NP_005017) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.1 EC 2.7.1.153 p110D P110DELTA Phosphatidylinositol-45-bisphosphate 3-kinase 110 kDa catalytic subunit delta phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit delta isoform phosphoinositide-3-kinase C phosphoinositide-3-kinase catalytic delta polypeptide PI3K PI3K-delta PI3-kinase p110 subunit delta PI3-kinase subunit delta PtdIns-3-kinase subunit delta PtdIns-3-kinase subunit p110-delta; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit delta; phosphoinositide-3-kinase C; phosphoinositide-3-kinase, catalytic, delta polypeptide variant p37delta; PI3-kinase p110 subunit delta; PI3-kinase subunit delta; PI3Kdelta; PtdIns-3-kinase subunit p110-delta
Gene Aliases: APDS; IMD14; p110D; P110DELTA; PI3K; PIK3CD
UniProt ID: (Human) O00329
Entrez Gene ID: (Human) 5293
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.