Novus Biologicals
Manufacturer Code:NBP299597FR
Catalog # NB085791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PI 3-Kinase p110 beta/PIK3CB. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: P110BETA PI3K PI3KBETA PIK3C1 PIK3CB phosphatidylinositol-45-bisphosphate 3-kinase catalytic subunit beta; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit beta isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta; phosphoinositide-3-kinase, catalytic, beta polypeptide; PI3-kinase p110 subunit beta; PI3-kinase subunit beta; PI3K-beta; PtdIns-3-kinase p110; ptdIns-3-kinase subunit beta; PtdIns-3-kinase subunit p110-beta
Gene Aliases: P110BETA; PI3K; PI3KBETA; PIK3C1; PIK3CB
UniProt ID: (Human) P42338
Entrez Gene ID: (Human) 5291
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.