Novus Biologicals
Manufacturer Code:NBP273395B
Catalog # NBP273395B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B1 |
Immunogen | Full length recombinant protein of human PI 3-Kinase C2 beta (NP_002637) produced in HEK293T cells. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: C2-PI3K; C2-PI3KDKFZp686G16234 EC 2.7.1 EC 2.7.1.154 phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta Phosphoinositide 3-kinase-C2-beta phosphoinositide-3-kinase class 2 beta polypeptide PI3K-C2beta PI3K-C2-beta PTDINS-3-kinase C2 beta PtdIns-3-kinase C2 subunit beta; phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide; Phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit beta; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 beta; Phosphoinositide 3-kinase-C2-beta; phosphoinositide-3-kinase, class 2, beta polypeptide; PI3K-C2-beta; PI3K-C2beta; PTDINS-3-kinase C2 beta; ptdIns-3-kinase C2 subunit beta
Gene Aliases: C2-PI3K; PIK3C2B
UniProt ID: (Human) O00750
Entrez Gene ID: (Human) 5287
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.