Proteintech
Manufacturer Code:161251AP150UL
Catalog # 50-173-0734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHYHD1 Fusion Protein Ag9108 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PHYHD1; Phytanoyl-CoA dioxygenase domain-containing protein 1
Gene Aliases: PHYHD1
UniProt ID: (Human) Q5SRE7, (Mouse) Q9DB26, (Rat) Q5BJP9
Entrez Gene ID: (Human) 254295, (Mouse) 227696, (Rat) 296621
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.