Novus Biologicals
Manufacturer Code:NBP290216B
Catalog # NB071056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human PHYH (O14832 Ser31-Leu338). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: LNAP1 PAHXLN1 peroxisomal PhyH PHYH1 phytanoyl-CoA 2-hydroxylase phytanoyl-CoA hydroxylase phytanoyl-CoA hydroxylase (Refsum disease) RD; PhyH; Phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; Phytanoyl-CoA alpha-hydroxylase; Phytanoyl-CoA dioxygenase, peroxisomal
Gene Aliases: LN1; LNAP1; PAHX; PHYH; PHYH1; RD
UniProt ID: (Human) O14832
Entrez Gene ID: (Human) 5264
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.