Invitrogen
Manufacturer Code:PA544595
Catalog # PIPA544595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PHYH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: induced by chronic shear stress; Lupus nephritis-associated peptide 1; PhyH; Phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; phytanoyl-CoA 2-hydroxylase; Phytanoyl-CoA alpha-hydroxylase; phytanoyl-CoA dioxygenase peroxisomal; Phytanoyl-CoA dioxygenase, peroxisomal; phytanoyl-CoA hydroxylase; phytanoyl-CoA hydroxylase (Refsum disease)
Gene Aliases: AI256161; AI265699; LN1; LNAP1; PAHX; PHYH; PHYH1; RD; zgc:110203
UniProt ID: (Bovine) O18778, (Human) O14832, (Mouse) O35386, (Rat) P57093
Entrez Gene ID: (Horse) 100056651, (Bovine) 281400, (Human) 5264, (Pig) 100134962, (Mouse) 16922, (Rat) 114209, (Guinea pig) 100721980, (Zebrafish) 550521
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.