Invitrogen
Manufacturer Code:PA526017
Catalog # PIPA526017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 59-87 amino acids from the N-terminal region of human PHYH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-HPCL HPCL HPCL2 PHYH2 1600020H07Rik 2-hydroxyphytanol-CoA lyase 2-hydroxyphytanoyl-CoA lyase phytanoyl-CoA 2-hydroxylase 2 phytanoyl-CoA hydroxylase 2; PhyH; Phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; Phytanoyl-CoA alpha-hydroxylase; Phytanoyl-CoA dioxygenase, peroxisomal
Gene Aliases: LN1; LNAP1; PAHX; PHYH; PHYH1; RD
UniProt ID: (Human) O14832
Entrez Gene ID: (Human) 5264
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.