Novus Biologicals
Manufacturer Code:NBP29010950UL
Catalog # NB021682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 7 |
Immunogen | Recombinant Human PHPT1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 14 kDa phosphohistidine phosphatase; bA216L13.10 DKFZp564M173 EC 3.1.314 kDa phosphohistidine phosphatase EC 3.1.3.- HSPC141 phosphohistidine phosphatase 11700008C22Rik phosphohistidine phosphatase 14kDa PHP14CGI-202 Protein janus-A homolog RP11-216L13.10 sex-regulated protein janus-a; epididymis secretory sperm binding protein Li 132P; Phosphohistidine phosphatase 1; PHP; PHPT1; Protein histidine phosphatase; Protein janus-A homolog; sex-regulated protein janus-a
Gene Aliases: CGI-202; HEL-S-132P; HSPC141; PHP14; PHPT1
UniProt ID: (Human) Q9NRX4
Entrez Gene ID: (Human) 29085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.