Proteintech
Manufacturer Code:131121AP150UL
Catalog # 50-172-8497
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHPT1 Fusion Protein Ag3778 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14 kDa phosphohistidine phosphatase; CGI 202 HSPC141 phosphohistidine phosphatase 1 PHP14 PHPT1 Protein janus A homolog; epididymis secretory sperm binding protein Li 132P; Phosphohistidine phosphatase 1; PHP; PHPT1; Protein histidine phosphatase; Protein janus-A homolog; sex-regulated protein janus-a
Gene Aliases: 1700008C22Rik; CGI-202; HEL-S-132P; HSPC141; PHP14; PHPT1
UniProt ID: (Human) Q9NRX4, (Mouse) Q9DAK9
Entrez Gene ID: (Human) 29085, (Mouse) 75454, (Rat) 296571
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.