Proteintech
Manufacturer Code:662541IG150UL
Catalog # 50-173-6687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F5F5 |
Immunogen | PHOX2B Fusion Protein Ag13196 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dilated pupils 1; GENA 269; NBPhox; NBPhox Neuroblastoma Phox paired like homeobox 2b PHOX2B PHOX2B homeodomain protein PMX2B; neuroblastoma paired-type homeobox protein; Neuroblastoma Phox; paired mesoderm homeobox 2b; Paired mesoderm homeobox protein 2B; Paired-like homeobox 2B; PHOX2B homeodomain protein
Gene Aliases: Dilp1; NBLST2; NBPhox; PHOX2B; PMX2B
UniProt ID: (Human) Q99453, (Mouse) O35690
Entrez Gene ID: (Human) 8929, (Mouse) 18935
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.