Invitrogen
Manufacturer Code:PA534381
Catalog # PIPA534381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid synthetic peptide near the carboxy terminus of human PHOX2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aristaless homeobox gene homolog; aristaless homeobox homolog; Aristaless homeobox protein homolog; arix homeodomain protein; ARIX1 homeodomain protein; paired mesoderm homeobox 2a; Paired mesoderm homeobox protein 2A; Paired-like homeobox 2A; PHOX2A homeodomain protein; PHOX2A Paired-like homeobox 2A Paired mesoderm homeobox protein 2A PMX2A NCAM2 FEOM2 CFEOM2 ARIX
Gene Aliases: ARIX; CFEOM2; FEOM2; NCAM2; Phox2; PHOX2A; Pmx2; PMX2A
UniProt ID: (Human) O14813, (Mouse) Q62066, (Rat) Q62782
Entrez Gene ID: (Human) 401, (Mouse) 11859, (Rat) 116648
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.