Invitrogen
Manufacturer Code:PA572134
Catalog # PIPA572134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 12-40 amino acids from the N-terminal region of human PHOSPHO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pyridoxal phosphate phosphatase PHOSPHO2; Pyridoxal phosphate phosphatase PHOSPHO2 PHOSPHO2
Gene Aliases: 1700048E23Rik; AI661517; AU021728; AV006103; Phos2; PHOSPHO2
UniProt ID: (Human) Q8TCD6, (Mouse) Q9D9M5
Entrez Gene ID: (Human) 493911, (Mouse) 73373
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.