Invitrogen
Manufacturer Code:PA576870
Catalog # PIPA576870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PHLDA2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beckwith Wiedemann syndrome chromosome region 1 candidate protein CBeckwith-Wiedemann syndrome chromosomal region 1 candidate gene C proteinBRW 1CBRW1CBWR 1CBWR1CHLDA 2HLDA2Imprinted in placenta and liverImprinted in placenta and liver proteinIPLIPLp17 Beckwith Wiedemann region 1Cp17 BWR1Cp17-Beckwith-Wiedemann region 1 Cp17-BWR1CPHLA2&lowbarHUMANPHLDA 2phlda2Pleckstrin homology like domain family A member 2Pleckstrin homology-like domain family A member 2TSSC 3Tumor suppressing STF cDNA 3 proteinTumor suppressing subchromosomal transferable fragment candidate gene 3 proteinTumor suppressing subchromosomal transferable fragment cDNA 3Tumor suppressing subtransferable candidate 3Tumor supressing STF cDNA 3Tumor-suppressing STF cDNA 3 proteinTumor-suppressing subchromosomal transferable fragment candidate gene 3 protein; Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene C protein; Imprinted in placenta and liver protein; p17-Beckwith-Wiedemann region 1 C; p17-Beckwith-Wiedemann region 1C; p17-BWR1C; Pleckstrin homology-like domain family A member 2; pleckstrin homology-like domain, family A, member 2; tumor suppressing subchromosomal transferable fragment cDNA 3; tumor suppressing subtransferable candidate 3; Tumor-suppressing STF cDNA 3 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 3 protein; tumor-supressing STF cDNA 3
Gene Aliases: BRW1C; BWR1C; HLDA2; IPL; PHLDA2; TSSC3
UniProt ID: (Human) Q53GA4
Entrez Gene ID: (Human) 7262
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.