Invitrogen
Manufacturer Code:PA570230
Catalog # PIPA570230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat PHKG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma phosphorylase kinase; PHK-gamma-M; PHKG phosphorylase b kinase gamma catalytic chain skeletal muscle isoform phosphorylase kinase gamma phosphorylase kinase subunit gamma-1; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase B kinase gamma catalytic chain, skeletal muscle isoform (Phosphorylase kinase gamma subunit 1); Phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase gamma; phosphorylase kinase gamma 1; phosphorylase kinase gamma subunit; phosphorylase kinase gamma subunit 1; phosphorylase kinase subunit gamma 1; Phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1 (muscle); Serine/threonine-protein kinase PHKG1; unnamed protein product
Gene Aliases: PHGK; PHKG; PHKG1; PHKIN01
UniProt ID: (Human) Q16816, (Rabbit) P00518, (Rat) P13286, (Mouse) P07934
Entrez Gene ID: (Human) 5260, (Bovine) 540682, (Rabbit) 100009297, (Horse) 100061404, (Pig) 397548, (Dog) 489784, (Goat) 102185139, (Guinea pig) 100732648, (Rat) 29353, (Mouse) 18682
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.