Invitrogen
Manufacturer Code:PA552902
Catalog # PIPA552902
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PHKG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PHK-gamma-M; PHKG phosphorylase b kinase gamma catalytic chain skeletal muscle isoform phosphorylase kinase gamma phosphorylase kinase subunit gamma-1; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase gamma; phosphorylase kinase gamma subunit 1; Phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1 (muscle); Serine/threonine-protein kinase PHKG1
Gene Aliases: PHKG; PHKG1
UniProt ID: (Human) Q16816
Entrez Gene ID: (Human) 5260
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.