Invitrogen
Manufacturer Code:PA5102028
Catalog # PIPA5102028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PHKG1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma phosphorylase kinase Phkg Phkg1 PHKgammaM PHK-gammaM PHK-gamma-M PHKgamma-M PHKIN01 phosphorylase b kinase gamma catalytic chain skeletal muscle isoform Phosphorylase B kinase gamma catalytic chain skeletal muscle isoform (Phosphorylase kinase gamma subunit 1) phosphorylase b kinase gamma catalytic chain skeletal muscle/heart isoform phosphorylase kinase phosphorylase kinase catalytic subunit gamma 1 phosphorylase kinase gamma phosphorylase kinase gamma 1 phosphorylase kinase gamma subunit 1 phosphorylase kinase subunit gamma 1 phosphorylase kinase subunit gamma-1 phosphorylase kinase gamma 1 phosphorylase kinase gamma 1 (muscle) serine/threonine-protein kinase PHKG1; PHK-gamma-M; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase gamma; phosphorylase kinase gamma subunit 1; Phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1 (muscle); Serine/threonine-protein kinase PHKG1
Gene Aliases: PHKG; PHKG1
UniProt ID: (Human) Q16816
Entrez Gene ID: (Human) 5260
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.