Abnova
Manufacturer Code:H00005257A01
Catalog # 89-105-104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 984 a.a. &bsim 1093 a.a) partial recombinant protein with GST tag.,PHKB (NP_000284 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PHKB.
For Research Use Only
Protein Aliases: DKFZp781E15103; FLJ41698; Phosphorylase b kinase regulatory subunit beta; phosphorylase kinase beta subunit; phosphorylase kinase beta-subunit; Phosphorylase kinase subunit beta; phosphorylase kinase, beta
Gene Aliases: PHKB
UniProt ID: (Human) Q93100
Entrez Gene ID: (Human) 5257
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.