Proteintech
Manufacturer Code:134001AP150UL
Catalog # 50-172-8713
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHKB Fusion Protein Ag4240 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PHKB phosphorylase kinase beta; Phosphorylase b kinase regulatory subunit beta; phosphorylase kinase beta subunit; phosphorylase kinase beta-subunit; Phosphorylase kinase subunit beta; phosphorylase kinase, beta
Gene Aliases: AI463271; PHKB
UniProt ID: (Human) Q93100, (Mouse) Q7TSH2
Entrez Gene ID: (Human) 5257, (Mouse) 102093
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.