Invitrogen
Manufacturer Code:PA521933
Catalog # PIPA521933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1172 and 1235 of PHKA2 (Uniprot IDNo.P46019) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: phosphorylase b kinase regulatory subunit alpha liver isoform; Phosphorylase b kinase regulatory subunit alpha, liver isoform; Phosphorylase kinase alpha L subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase, alpha 2 (liver); RP3-499B10.2 GSD9A PHK PYK PYKL XLG XLG2 phosphorylase b kinase regulatory subunit alpha liver isoform phosphorylase b kinase regulatory subunit alpha liver isoform phosphorylase kinase alpha L subunit phosphorylase kinase alpha-subunit
Gene Aliases: GSD9A; PHK; PHKA2; PHKLA; PYK; PYKL; XLG; XLG2
UniProt ID: (Human) P46019
Entrez Gene ID: (Human) 5256
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.