Novus Biologicals
Manufacturer Code:NBP192264
Catalog # NBP192264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GARVKLGNLSEFLTTSFYTYLTFLDPDCDEKLFDNASEGTFSPDSDSDLVGYLEDTCNQESQDELDHYINHLLQSTSLRSYLPPLCKNTEDRHVFSAIHSTRDILSVMAKAKGLEVPFVPMTLPTKVLSAHRKS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GSD9A MGC133071 PHK PHKLA phosphorylase b kinase regulatory subunit alpha liver isoform phosphorylase b kinase regulatory subunit alpha liver isoform Phosphorylase kinase alpha L subunit phosphorylase kinase alpha-subunit phosphorylase kinase alpha 2 (liver) PYK PYKL XLG XLG2; phosphorylase b kinase regulatory subunit alpha liver isoform; Phosphorylase b kinase regulatory subunit alpha, liver isoform; Phosphorylase kinase alpha L subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase, alpha 2 (liver)
Gene Aliases: GSD9A; PHK; PHKA2; PHKLA; PYK; PYKL; XLG; XLG2
UniProt ID: (Human) P46019
Entrez Gene ID: (Human) 5256
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.