Abnova
Manufacturer Code:H00005256A01
Catalog # 89-105-103
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 428 a.a. &bsim 521 a.a) partial recombinant protein with GST tag.,PHKA2 (NP_000283 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PHKA2.
For Research Use Only
Protein Aliases: GSD9A; MGC133071; PHK; phosphorylase b kinase regulatory subunit alpha liver isoform; Phosphorylase b kinase regulatory subunit alpha, liver isoform; Phosphorylase kinase alpha L subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase, alpha 2 (liver); PYK; PYKL; XLG; XLG2
Gene Aliases: GSD9A; PHK; PHKA2; PHKLA; PYK; PYKL; XLG; XLG2
UniProt ID: (Human) P46019
Entrez Gene ID: (Human) 5256
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.