Proteintech
Manufacturer Code:246581AP150UL
Catalog # 50-173-4476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHKA2 Fusion Protein Ag9473 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GSD9A PHK PHKA2 PHKLA PYK PYKL XLG XLG2; phosphorylase b kinase regulatory subunit alpha liver isoform; Phosphorylase b kinase regulatory subunit alpha, liver isoform; Phosphorylase kinase alpha L subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase, alpha 2 (liver)
Gene Aliases: 6330505C01Rik; D330034O08; GSD9A; PHK; PHKA2; PHKLA; PYK; PYKL; XLG; XLG2
UniProt ID: (Human) P46019, (Mouse) Q8BWJ3
Entrez Gene ID: (Human) 5256, (Mouse) 110094, (Rat) 678739
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.