Invitrogen
Manufacturer Code:PA599685
Catalog # PIPA599685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PHKA1/2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5330411D17 6330505C01Rik 9830108K24Rik D330034O08 GSD9A Pcyt1b PHK PHKA PHKA1 PHKA2 PhK-alpha-subunit PHKLA phosphorylase B kinase alpha subunit phosphorylase b kinase regulatory subunit alpha liver isoform phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform phosphorylase b kinase regulatory subunit alpha liver isoform Phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform phosphorylase kinase phosphorylase kinase alpha 1 phosphorylase kinase alpha 2 phosphorylase kinase alpha L subunit Phosphorylase kinase alpha M subunit phosphorylase kinase alpha-subunit phosphorylase kinase regulatory subunit alpha 1 phosphorylase kinase regulatory subunit alpha 2 phosphorylase kinase alpha 1 phosphorylase kinase alpha 1 (muscle) phosphorylase kinase alpha 1 (muscle) muscle glycogenosis phosphorylase kinase alpha 2 phosphorylase kinase alpha 2 (liver) PYK PYKL XLG XLG2; PhK-alpha-subunit; phosphorylase B kinase alpha subunit; phosphorylase b kinase regulatory subunit alpha liver isoform; phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform; Phosphorylase b kinase regulatory subunit alpha, liver isoform; Phosphorylase b kinase regulatory subunit alpha, skeletal muscle isoform; Phosphorylase kinase alpha L subunit; Phosphorylase kinase alpha M subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase, alpha 1 (muscle); phosphorylase kinase, alpha 1 (muscle), muscle glycogenosis; phosphorylase kinase, alpha 2 (liver)
Gene Aliases: 5330411D17; 6330505C01Rik; 9830108K24Rik; D330034O08; GSD9A; Pcyt1b; PHK; PHKA; PHKA1; PHKA2; PHKLA; PYK; PYKL; XLG; XLG2
UniProt ID: (Human) P46020, (Human) P46019, (Mouse) P18826, (Mouse) Q8BWJ3
Entrez Gene ID: (Human) 5255, (Human) 5256, (Rat) 64561, (Mouse) 18679, (Mouse) 110094, (Rat) 678739
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.