Novus Biologicals
Manufacturer Code:NBP187281
Catalog # NBP187281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AHSLRCSAEEATEGLMNLSPSAMKNLLHHILSGKEFGVERSVRPTDSNVSPAISIHEIGAVGATKTERTGIMQLKSEIKQSPGTSMTPSSGSFPSAYDQQSSKDSRQGQWQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC132604 PHKA phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform Phosphorylase kinase alpha M subunit phosphorylase kinase alpha 1 (muscle) phosphorylase kinase alpha 1 (muscle) muscle glycogenosis; phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform; Phosphorylase b kinase regulatory subunit alpha, skeletal muscle isoform; Phosphorylase kinase alpha M subunit; phosphorylase kinase, alpha 1 (muscle); phosphorylase kinase, alpha 1 (muscle), muscle glycogenosis
Gene Aliases: PHKA; PHKA1
UniProt ID: (Human) P46020
Entrez Gene ID: (Human) 5255
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.