Proteintech
Manufacturer Code:242791AP150UL
Catalog # 50-173-4272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHKA1 Fusion Protein Ag17998 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PhK-alpha-subunit; PHKA PHKA1; phosphorylase B kinase alpha subunit; phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform; Phosphorylase b kinase regulatory subunit alpha, skeletal muscle isoform; Phosphorylase kinase alpha M subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase, alpha 1 (muscle); phosphorylase kinase, alpha 1 (muscle), muscle glycogenosis
Gene Aliases: 5330411D17; 9830108K24Rik; Pcyt1b; PHKA; PHKA1
UniProt ID: (Human) P46020, (Mouse) P18826
Entrez Gene ID: (Human) 5255, (Rat) 64561, (Mouse) 18679
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.