Proteintech
Manufacturer Code:209331AP150UL
Catalog # 50-173-3162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHIP Fusion Protein Ag14968 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDB1 and CUL4 associated factor 14; DDB1- and CUL4-associated factor 14; IRS-1 PH domain-binding protein; ndrp PH interacting protein PHIP WDR11; neuronal differentiation related protein; neuronal differentiation-related protein; PH-interacting protein; PHIP; WD repeat domain 11; WD repeat-containing protein 11
Gene Aliases: 2810004D21Rik; 4632404O06Rik; BRWD2; DCAF14; Ndrp; PHIP; WDR11
UniProt ID: (Human) A7J992
Entrez Gene ID: (Human) 55023, (Mouse) 83946
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.