Abnova
Manufacturer Code:PAB14120
Catalog # 89-116-940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A mixture of synthetic peptides (conjugated with KLH) that corresponding to human PHF8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PHF8.
For Research Use Only
Protein Aliases: [histone H3]-dimethyl-L-lysine(36) demethylase PHF8; [histone H3]-dimethyl-L-lysine(9) demethylase PHF8; DKFZp686E0868; Histone lysine demethylase PHF8; JHDM1F; jumonji C domain-containing histone demethylase 1F; KIAA1111; MRXSSD; PHD finger protein 8; ZNF422
Gene Aliases: JHDM1F; KIAA1111; MRXSSD; PHF8; ZNF422
UniProt ID: (Human) Q9UPP1
Entrez Gene ID: (Human) 23133
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.