Novus Biologicals
Manufacturer Code:NBP281980
Catalog # NB017291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHF14 antibody was raised against a 19 amino acid peptide near the amino terminus of human PHF14. The immunogen is located within amino acids 210 - 260 of PHF14 . Amino Acid Squence: KRKGRSASQKEGSDGDNED |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0783 MGC176640 PHD finger protein 14; PHD finger protein 14
Gene Aliases: KIAA0783; PHF14
UniProt ID: (Human) O94880
Entrez Gene ID: (Human) 9678
Molecular Function: nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.