Invitrogen
Manufacturer Code:PA540637
Catalog # PIPA540637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human PHF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPCl1; mPCl1; PHD finger protein 1; PHD finger protein 1 PCL1 PHF2 MTF2L2; polycomb-like 1; Polycomb-like protein 1; Protein PHF1; T-complex testis-expressed 3; testicular tissue protein Li 140; tudor domain containing 19C
Gene Aliases: AW557215; D17Ertd455e; hPHF1; MTF2L2; PCL1; PHF1; PHF2; Plc1; Tctex-3; Tctex3; TDRD19C
UniProt ID: (Human) O43189, (Mouse) Q9Z1B8
Entrez Gene ID: (Bovine) 508957, (Human) 5252, (Dog) 481741, (Rabbit) 100346719, (Horse) 100061619, (Guinea pig) 100729738, (Mouse) 21652, (Rat) 294287
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.