Novus Biologicals
Manufacturer Code:NBP298955IR
Catalog # NB106934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PHF1. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPCl1; hPCl1 MTF2L2 PCL1 PHD finger protein 1 PHF2 Polycomb-like protein 1 Protein PHF1; PHD finger protein 1; Polycomb-like protein 1; Protein PHF1; testicular tissue protein Li 140; tudor domain containing 19C
Gene Aliases: hPHF1; MTF2L2; PCL1; PHF1; PHF2; TDRD19C
UniProt ID: (Human) O43189
Entrez Gene ID: (Human) 5252
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.