Proteintech
Manufacturer Code:156631AP150UL
Catalog # 50-173-0386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PHF1 Fusion Protein Ag8094 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPCl1; hPCl1 MTF2L2 PCL1 PHD finger protein 1 PHF1 PHF2 Polycomb like protein 1 Protein PHF1; mPCl1; PHD finger protein 1; polycomb-like 1; Polycomb-like protein 1; Protein PHF1; T-complex testis-expressed 3; testicular tissue protein Li 140; tudor domain containing 19C
Gene Aliases: AW557215; D17Ertd455e; hPHF1; MTF2L2; PCL1; PHF1; PHF2; Plc1; Tctex-3; Tctex3; TDRD19C
UniProt ID: (Human) O43189, (Mouse) Q9Z1B8
Entrez Gene ID: (Human) 5252, (Mouse) 21652, (Rat) 294287
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.