Invitrogen
Manufacturer Code:MA516144
Catalog # PIMA516144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | EG188e/d5 |
Immunogen | Partial recombit human PHD3 protein (UniProt accession number Q9H6Z9) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Egl nine homolog 3; egl nine homolog 3, mitochondrial; egl nine-like protein 3 isoform; egl-9 family hypoxia-inducible factor 3; factor-responsive smooth muscle protein; HIF prolyl hydroxylase 3; HIF-PH3; HIF-prolyl hydroxylase 3; HIFPH3 PHD3 HIF prolyl hydroxylase 3 HIF-PH3 HIF-prolyl hydroxylase 3 HPH-1 HPH-3 egl nine homolog 3 egl nine-like protein 3 isoform hypoxia-inducible factor prolyl hydroxylase 3 prolyl hydroxylase domain-containing protein 3; HPH-1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 3; PHD3; Prolyl hydroxylase domain-containing protein 3; Prolyl hydroxylase EGLN3; SM-20
Gene Aliases: EGLN3; HIFP4H3; HIFPH3; PHD-3; PHD3; SM-20; Sm20
UniProt ID: (Human) Q9H6Z9, (Rat) Q62630
Entrez Gene ID: (Human) 112399, (Rat) 54702
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.