Invitrogen
Manufacturer Code:PA5100300
Catalog # PIPA5100300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PHD3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610021G09Rik AI505553 AI648162 Egl nine homolog 3 egl nine homolog 3 mitochondrial egl nine-like protein 3 isoform egl-9 family hypoxia inducible factor 3 egl-9 family hypoxia-inducible factor 3 EGLN3 factor-responsive smooth muscle protein FLJ21620 HIF prolyl hydroxylase 3 HIFP4H3 Hif-p4h-3 HIFPH3 HIF-PH3 HIF-prolyl hydroxylase 3 HPH-1 HPH-3 Hypoxia-inducible factor prolyl hydroxylase 3 MGC125998 MGC125999 P4H3 PHD3 PHD-3 Prolyl hydroxylase domain-containing protein 3 Sm20 SM-20; Egl nine homolog 3; egl nine-like protein 3 isoform; egl-9 family hypoxia-inducible factor 3; HIF prolyl hydroxylase 3; HIF-PH3; HIF-prolyl hydroxylase 3; HPH-1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 3; PHD3; Prolyl hydroxylase domain-containing protein 3; Prolyl hydroxylase EGLN3; SM-20
Gene Aliases: 2610021G09Rik; AI505553; AI648162; EGLN3; Hif-p4h-3; HIFP4H3; HIFPH3; PHD3; SM-20
UniProt ID: (Human) Q9H6Z9, (Mouse) Q8CCA8
Entrez Gene ID: (Human) 112399, (Mouse) 112407
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.