Invitrogen
Manufacturer Code:PA116526
Catalog # APPA6526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide containing residues between 50-100 of human PHD3/HIF Prolyl Hydroxylase 3 using the numbering given in entry NP&lowbar0713561 (GeneID 112399) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Egl nine homolog 3; egl nine homolog 3, mitochondrial; egl nine-like protein 3 isoform; egl-9 family hypoxia-inducible factor 3; EGL9 homolog of C. elegans 3 EGLN3 Factor responsive smooth muscle protein HIF Prolyl Hydroxylase 3 HIFP4H3 HIFPH3 P4H3 Prolyl Hydroxylase Doma; factor-responsive smooth muscle protein; HIF prolyl hydroxylase 3; HIF-PH3; HIF-prolyl hydroxylase 3; HPH-1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 3; PHD3; Prolyl hydroxylase domain-containing protein 3; Prolyl hydroxylase EGLN3; SM-20
Gene Aliases: 2610021G09Rik; AI505553; AI648162; EGLN3; Hif-p4h-3; HIFP4H3; HIFPH3; PHD-3; PHD3; SM-20; Sm20
UniProt ID: (Human) Q9H6Z9, (Mouse) Q8CCA8, (Rat) Q62630
Entrez Gene ID: (Human) 112399, (Mouse) 112407, (Rat) 54702
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.