Invitrogen
Manufacturer Code:MA516143
Catalog # PIMA516143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 366G/76/3 |
Immunogen | A peptide within residues 1-50 of the human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia-inducible factor 1; HIF prolyl hydroxylase 2; HIF-PH2; HIF-prolyl hydroxylase 2; Hypoxia-inducible factor prolyl hydroxylase 2; PHD2; PNAS-118 C1orf12 ECYT3 HIF-PH2 HIFPH2 HPH-2 HPH2 PHD2 SM20 ZMYND6 HIF prolyl hydroxylase 2 HIF-prolyl hydroxylase 2 egl nine homolog 1 egl nine-like protein 1 hypoxia-inducible factor prolyl hydroxylase 2 prolyl hydroxylase domain-containing protein 2 zinc finger MYND domain-containing protein 6; Prolyl hydroxylase domain-containing protein 2; SM-20; zinc finger MYND domain-containing protein 6
Gene Aliases: C1orf12; ECYT3; EGLN1; HALAH; HIF-PH2; HIFPH2; HPH-2; HPH2; PHD2; PNAS-118; PNAS-137; SM20; ZMYND6
UniProt ID: (Human) Q9GZT9
Entrez Gene ID: (Human) 54583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.