Invitrogen
Manufacturer Code:PA578511
Catalog # PIPA578511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PHD2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf12 ECYT3 EGLN1 HIF PH2 HIFPH2 HPH2 SM20 ZMYND6 egl-9 family hypoxia-inducible factor 1; Egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia-inducible factor 1; HIF prolyl hydroxylase 2; HIF-PH2; HIF-prolyl hydroxylase 2; Hypoxia-inducible factor prolyl hydroxylase 2; PHD2; Prolyl hydroxylase domain-containing protein 2; SM-20; zinc finger MYND domain-containing protein 6
Gene Aliases: AI503754; C1orf12; ECYT3; EGLN1; HALAH; Hif-p4h-2; HIF-PH2; HIFPH2; HPH-2; HPH2; ORF13; PHD2; PNAS-118; PNAS-137; SM-20; SM20; ZMYND6
UniProt ID: (Human) Q9GZT9, (Mouse) Q91YE3
Entrez Gene ID: (Human) 54583, (Mouse) 112405
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.